missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybiphenyl 96.0+%, TCI America™

Catalog No. M13585G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M13585G Supplier TCI America Supplier No. M13585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2113-56-6
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.24
MDL Number MFCD00027514
InChI Key KQMIWCAOEFUBQK-UHFFFAOYSA-N
PubChem CID 257971
IUPAC Name 3-methoxy-1,1'-biphenyl
SMILES COC1=CC=CC(=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 184.24
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxybiphenyl