missing translation for 'onlineSavingsMsg'
Learn More

3'-Methoxybiphenyl-4-sulfonyl chloride, 95%, Thermo Scientific™

Chemical Identifiers

CAS 186550-26-5
Molecular Formula C13H11ClO3S
Molecular Weight (g/mol) 282.75
InChI Key DVQBUJTYQXKLKZ-UHFFFAOYSA-N
Synonym 3'-methoxy-biphenyl-4-sulfonyl chloride, 3'-methoxybiphenyl-4-sulfonyl chloride, 3'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 3'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-methoxy, 4-3-methoxyphenyl benzenesulfonyl chloride, 3'-methoxy-biphenyl-4-sulfonylchloride, 3/'-methoxybiphenyl-4-sulfonyl chloride
PubChem CID 10826798
IUPAC Name 4-(3-methoxyphenyl)benzenesulfonyl chloride
SMILES COC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC450480050
SDS View Documents
Thermo Scientific Chemicals
450480050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

186550-26-5
282.75
3'-methoxy-biphenyl-4-sulfonyl chloride, 3'-methoxybiphenyl-4-sulfonyl chloride, 3'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 3'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-methoxy, 4-3-methoxyphenyl benzenesulfonyl chloride, 3'-methoxy-biphenyl-4-sulfonylchloride, 3/'-methoxybiphenyl-4-sulfonyl chloride
4-(3-methoxyphenyl)benzenesulfonyl chloride
C13H11ClO3S
DVQBUJTYQXKLKZ-UHFFFAOYSA-N
10826798
COC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

186550-26-5
C13H11ClO3S
3'-methoxy-biphenyl-4-sulfonyl chloride, 3'-methoxybiphenyl-4-sulfonyl chloride, 3'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 3'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-methoxy, 4-3-methoxyphenyl benzenesulfonyl chloride, 3'-methoxy-biphenyl-4-sulfonylchloride, 3/'-methoxybiphenyl-4-sulfonyl chloride
COC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
282.75
282.75
3'-Methoxybiphenyl-4-sulfonyl chloride
94% min. (HPLC)
5g
DVQBUJTYQXKLKZ-UHFFFAOYSA-N
4-(3-methoxyphenyl)benzenesulfonyl chloride
10826798
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only