missing translation for 'onlineSavingsMsg'
Learn More

(3-Methoxybenzyl)triphenylphosphonium Chloride 98.0+%, TCI America™

Catalog No. M24631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24631G 1 g
M24635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24631G Supplier TCI America Supplier No. M24631G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18880-05-2
Molecular Formula C26H24ClOP
Molecular Weight (g/mol) 418.901
MDL Number MFCD09836189
InChI Key DPYDLIVUYPUXBV-UHFFFAOYSA-M
PubChem CID 11407464
IUPAC Name (3-methoxyphenyl)methyl-triphenylphosphanium;chloride
SMILES COC1=CC=CC(=C1)C[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Cl-]

Specifications

Melting Point 268°C
Color White
Quantity 1 g
Formula Weight 418.90
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (3-Methoxybenzyl)triphenylphosphonium Chloride