missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxybenzonitrile 95.0+%, TCI America™
$48.13 - $136.69
Chemical Identifiers
| CAS | 1527-89-5 |
|---|---|
| Molecular Formula | C8H7NO |
| Molecular Weight (g/mol) | 133.15 |
| MDL Number | MFCD00001801 |
| InChI Key | KLXSUMLEPNAZFK-UHFFFAOYSA-N |
| Synonym | m-cyanoanisole, benzonitrile, 3-methoxy, m-anisonitrile, m-methoxybenzonitrile, 3-cyanoanisole, 3-anisonitrile, benzonitrile, m-methoxy, 3-methoxy-benzonitrile, 3-methoxy benzonitrile, m-methoxybenzontrile |
| PubChem CID | 73712 |
| IUPAC Name | 3-methoxybenzonitrile |
| SMILES | COC1=CC=CC(=C1)C#N |
Chemical Identifiers
| 1527-89-5 | |
| 133.15 | |
| KLXSUMLEPNAZFK-UHFFFAOYSA-N | |
| 73712 | |
| COC1=CC=CC(=C1)C#N |
| C8H7NO | |
| MFCD00001801 | |
| m-cyanoanisole, benzonitrile, 3-methoxy, m-anisonitrile, m-methoxybenzonitrile, 3-cyanoanisole, 3-anisonitrile, benzonitrile, m-methoxy, 3-methoxy-benzonitrile, 3-methoxy benzonitrile, m-methoxybenzontrile | |
| 3-methoxybenzonitrile |
Specifications
| 1527-89-5 | |
| 140°C | |
| MFCD00001801 | |
| 3439 | |
| KLXSUMLEPNAZFK-UHFFFAOYSA-N | |
| 3-methoxybenzonitrile | |
| 73712 | |
| ≥95.0% (GC) | |
| 3-Methoxybenzonitrile |
| White-Yellow | |
| C8H7NO | |
| 5 g | |
| m-cyanoanisole, benzonitrile, 3-methoxy, m-anisonitrile, m-methoxybenzonitrile, 3-cyanoanisole, 3-anisonitrile, benzonitrile, m-methoxy, 3-methoxy-benzonitrile, 3-methoxy benzonitrile, m-methoxybenzontrile | |
| COC1=CC=CC(=C1)C#N | |
| 133.15 | |
| 133.15 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No