missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzonitrile 95.0+%, TCI America™

Catalog No. M12915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12915G 5 g
M129125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12915G Supplier TCI America Supplier No. M12915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1527-89-5
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00001801
InChI Key KLXSUMLEPNAZFK-UHFFFAOYSA-N
Synonym m-cyanoanisole, benzonitrile, 3-methoxy, m-anisonitrile, m-methoxybenzonitrile, 3-cyanoanisole, 3-anisonitrile, benzonitrile, m-methoxy, 3-methoxy-benzonitrile, 3-methoxy benzonitrile, m-methoxybenzontrile
PubChem CID 73712
IUPAC Name 3-methoxybenzonitrile
SMILES COC1=CC=CC(=C1)C#N

Specifications

Color White-Yellow
Boiling Point 140°C
Quantity 5 g
UN Number 3439
Formula Weight 133.15
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Methoxybenzonitrile