Learn More
3-Methoxybenzhydrazide, 98+%
CAS: 5785-06-8 | C8H10N2O2 | 166.18 g/mol
Supplier: Thermo Scientific Chemicals B2095614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Methoxybenzhydrazide | |
| 90°C to 94°C | |
| MFCD00007601 | |
| 2092135 | |
| VMZSDAQEWPNOIB-UHFFFAOYSA-N | |
| 3-methoxybenzohydrazide | |
| 79848 | |
| ≥98% |
| 5785-06-8 | |
| C8H10N2O2 | |
| 25 g | |
| 3-methoxybenzhydrazide, m-anisic hydrazide, m-anisohydrazide, m-methoxy benzhydrazide, 3-methoxybenzoic hydrazide, benzoylhydrazine, m-methoxy, 3-methoxybenzenecarbohydrazide, 3-methoxybenzene-1-carbohydrazide, benzoic acid, 3-methoxy-, hydrazide, m-anisoylhydrazine | |
| COC1=CC=CC(=C1)C(=O)NN | |
| 166.18 | |
| 166.18 |
Chemical Identifiers
| 5785-06-8 | |
| MFCD00007601 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 227-310-0
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only