missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzhydrazide, 98+%

Catalog No. p-7025473
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2095606 5 g
AAB2095614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2095606 Supplier Thermo Scientific Chemicals Supplier No. B2095606

CAS: 5785-06-8 | C8H10N2O2 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5785-06-8
MDL Number MFCD00007601

Specifications

Melting Point 90°C to 94°C
Molecular Formula C10H16
Quantity 5 g
Beilstein 2092135
Synonym 1s-2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, s-camphene, 1s,4r-2,2-dimethyl-3-methylene-bicyclo 2.2.1 heptane
InChI Key CRPUJAZIXJMDBK-UHFFFAOYNA-N
SMILES CC1(C2CCC(C2)C1=C)C
IUPAC Name (1S,4S)-3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane
Molecular Weight (g/mol) 136.24
PubChem CID 6560154
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 3-Methoxybenzhydrazide
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-310-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only