missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzenesulfonamide, 97%, Thermo Scientific Chemicals

Catalog No. AAH2884403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2884403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2884403 Supplier Thermo Scientific Chemicals Supplier No. H2884403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58734-57-9 | Molecular Formula: C7H9NO3S | Molecular Weight: 187.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58734-57-9
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.21
MDL Number MFCD08704579
InChI Key VBKIEQKVSHDVGH-UHFFFAOYSA-N
PubChem CID 14763060
IUPAC Name 3-methoxybenzenesulfonamide
SMILES COC1=CC(=CC=C1)S(N)(=O)=O

Specifications

Melting Point 131°C to 133°C
Quantity 1 g
Synonym 3-methoxybenzenesulphonamide, 3-methoxybenzene-1-sulfonamide, benzenesulfonamide, 3-methoxy, benzenesulfonamide,3-methoxy, 3-sulphamoylanisole, acmc-20alqa, m-methoxybenzensulfonamid, 3-aminosulphonyl anisole, benzenesulfonamide, 3-methoxy-9ci
Formula Weight 187.22
Percent Purity 97%
Chemical Name or Material 3-Methoxybenzenesulfonamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only