missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Methoxyacetophenone 97.0+%, TCI America™
$78.29 - $577.86
Chemical Identifiers
| CAS | 586-37-8 |
|---|---|
| Molecular Formula | C9H10O2 |
| Molecular Weight (g/mol) | 150.177 |
| MDL Number | MFCD00008736 |
| InChI Key | BAYUSCHCCGXLAY-UHFFFAOYSA-N |
| Synonym | 3'-methoxyacetophenone, 3-methoxyacetophenone, 3-acetylanisole, m-methoxyacetophenone, 1-3-methoxyphenyl ethanone, ethanone, 1-3-methoxyphenyl, m-acetanisole, acetophenone, m-methoxy, 1-3-methoxyphenyl ethan-1-one, acetophenone, 3'-methoxy |
| PubChem CID | 11460 |
| IUPAC Name | 1-(3-methoxyphenyl)ethanone |
| SMILES | CC(=O)C1=CC(=CC=C1)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M010425ML
|
TCI America
M010425ML |
25 mL |
Each for $78.29
|
|
|||||
|
M0104500ML
|
TCI America
M0104500ML |
500 mL |
Each for $577.86
|
|
|||||
Chemical Identifiers
| 586-37-8 | |
| 150.177 | |
| BAYUSCHCCGXLAY-UHFFFAOYSA-N | |
| 11460 | |
| CC(=O)C1=CC(=CC=C1)OC |
| C9H10O2 | |
| MFCD00008736 | |
| 3'-methoxyacetophenone, 3-methoxyacetophenone, 3-acetylanisole, m-methoxyacetophenone, 1-3-methoxyphenyl ethanone, ethanone, 1-3-methoxyphenyl, m-acetanisole, acetophenone, m-methoxy, 1-3-methoxyphenyl ethan-1-one, acetophenone, 3'-methoxy | |
| 1-(3-methoxyphenyl)ethanone |
Specifications
| 586-37-8 | |
| 240°C | |
| MFCD00008736 | |
| 3'-methoxyacetophenone, 3-methoxyacetophenone, 3-acetylanisole, m-methoxyacetophenone, 1-3-methoxyphenyl ethanone, ethanone, 1-3-methoxyphenyl, m-acetanisole, acetophenone, m-methoxy, 1-3-methoxyphenyl ethan-1-one, acetophenone, 3'-methoxy | |
| CC(=O)C1=CC(=CC=C1)OC | |
| 150.177 | |
| 150.18 | |
| Liquid |
| Yellow | |
| C9H10O2 | |
| 25 mL | |
| BAYUSCHCCGXLAY-UHFFFAOYSA-N | |
| 1-(3-methoxyphenyl)ethanone | |
| 11460 | |
| ≥97.0% (GC) | |
| 3′-Methoxyacetophenone |
Safety and Handling
EINECSNumber : (3)-2676
TSCA : No