accessibility menu, dialog, popup

UserName

CH3OCH3O

CAS RN 586-37-8

IUPAC Name: 1-(3-methoxyphenyl)ethan-1-one
Synonyms: M-methoxyacetophenone
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: BAYUSCHCCGXLAY-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C(C)=O
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)