missing translation for 'onlineSavingsMsg'
Learn More

3'-Methoxyacetophenone 97.0+%, TCI America™

Catalog No. M010425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M010425ML 25 mL
M0104500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M010425ML Supplier TCI America Supplier No. M010425ML

Chemical Identifiers

CAS 586-37-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008736
InChI Key BAYUSCHCCGXLAY-UHFFFAOYSA-N
Synonym 3'-methoxyacetophenone, 3-methoxyacetophenone, 3-acetylanisole, m-methoxyacetophenone, 1-3-methoxyphenyl ethanone, ethanone, 1-3-methoxyphenyl, m-acetanisole, acetophenone, m-methoxy, 1-3-methoxyphenyl ethan-1-one, acetophenone, 3'-methoxy
PubChem CID 11460
IUPAC Name 1-(3-methoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)OC

Specifications

Color Yellow
Boiling Point 240°C
Quantity 25 mL
Formula Weight 150.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3′-Methoxyacetophenone
EINECSNumber (3)-2676
TSCA No