missing translation for 'onlineSavingsMsg'
Learn More

3'-Methoxyacetophenone, 97%

CAS: 586-37-8 | C9H10O2 | 150.177 g/mol

Supplier:  Thermo Scientific Chemicals A1560422

 View more versions of this product

Catalog No. AAA1560422

Only null left
Explore available promotions

Description

Description

It is applied in the asymmetric transfer hydrogenation of ketones catalyzed by ruthenium(II) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in the asymmetric transfer hydrogenation of ketones catalyzed by ruthenium(II) complexes.

Solubility
Fully miscible in water. Soluble in alcohol.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

3'-Methoxyacetophenone
-7°C
239°C to 241°C
C9H10O2
MFCD00008736
878552
3'-methoxyacetophenone, 3-methoxyacetophenone, 3-acetylanisole, m-methoxyacetophenone, 1-3-methoxyphenyl ethanone, ethanone, 1-3-methoxyphenyl, m-acetanisole, acetophenone, m-methoxy, 1-3-methoxyphenyl ethan-1-one, acetophenone, 3'-methoxy
BAYUSCHCCGXLAY-UHFFFAOYSA-N
1-(3-methoxyphenyl)ethanone
11460
97%
586-37-8
1.094
>110°C (230°F)
1.5425
100 g
Light sensitive
Fully miscible in water. Soluble in alcohol.
CC(=O)C1=CC(=CC=C1)OC
150.177
150.18

Chemical Identifiers

586-37-8
150.177
BAYUSCHCCGXLAY-UHFFFAOYSA-N
11460
CC(=O)C1=CC(=CC=C1)OC
C9H10O2
MFCD00008736
3'-methoxyacetophenone, 3-methoxyacetophenone, 3-acetylanisole, m-methoxyacetophenone, 1-3-methoxyphenyl ethanone, ethanone, 1-3-methoxyphenyl, m-acetanisole, acetophenone, m-methoxy, 1-3-methoxyphenyl ethan-1-one, acetophenone, 3'-methoxy
1-(3-methoxyphenyl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H312-H315-H319-H332

EINECSNumber : 209-573-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only