missing translation for 'onlineSavingsMsg'
Learn More

3'-Methoxyacetophenone, 97%

Catalog No. AAA1560436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1560436 500 g
AAA1560414 25 g
AAA1560422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1560436 Supplier Thermo Scientific Chemicals Supplier No. A1560436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 586-37-8 | C9H10O2 | 150.177 g/mol

It is applied in the asymmetric transfer hydrogenation of ketones catalyzed by ruthenium(II) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in the asymmetric transfer hydrogenation of ketones catalyzed by ruthenium(II) complexes.

Solubility
Fully miscible in water. Soluble in alcohol.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 586-37-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008736
InChI Key BAYUSCHCCGXLAY-UHFFFAOYSA-N
Synonym 3'-methoxyacetophenone, 3-methoxyacetophenone, 3-acetylanisole, m-methoxyacetophenone, 1-3-methoxyphenyl ethanone, ethanone, 1-3-methoxyphenyl, m-acetanisole, acetophenone, m-methoxy, 1-3-methoxyphenyl ethan-1-one, acetophenone, 3'-methoxy
PubChem CID 11460
IUPAC Name 1-(3-methoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)OC

Specifications

Melting Point -7°C
Density 1.094
Boiling Point 239°C to 241°C
Flash Point >110°C (230°F)
Refractive Index 1.5425
Quantity 500 g
Beilstein 878552
Sensitivity Light sensitive
Solubility Information Fully miscible in water. Soluble in alcohol.
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material 3'-Methoxyacetophenone
Show More Show Less
Hazard Category H302-H312-H315-H319-H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 209-573-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only