Learn More
3-Methoxy-o-phenylenediamine, 97%
CAS: 37466-89-0 | C7H10N2O | 138.17 g/mol
Supplier: Thermo Scientific Chemicals H3528603
Description
3-Methoxy-o-phenylenediamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methoxy-o-phenylenediamine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store away from oxidizing agents.
Specifications
| 3-Methoxy-o-phenylenediamine | |
| 39°C to 40°C | |
| MFCD08276903 | |
| UN2811 | |
| 2,3-diaminoanisole, 2,3-diaminoanisol, diaminoanisole, 3-methoxy-benzene-1,2-diamine, 3-methoxy-1,2-phenylenediamine, 1,2-benzenediamine, 3-methoxy, 3-methoxy-o-phenylenediamine, 3-methoxy-1,2-benzenediamine, ambkkkkk251, 2-amino-3-methoxyaniline | |
| BFLWXPJTAKXXKT-UHFFFAOYSA-N | |
| 3-methoxybenzene-1,2-diamine | |
| 184268 | |
| 97% |
| 37466-89-0 | |
| C7H10N2O | |
| 1 g | |
| Air sensitive | |
| Slightly soluble in water. | |
| COC1=CC=CC(=C1N)N | |
| 138.17 | |
| 138.17 |
Chemical Identifiers
| 37466-89-0 | |
| 138.17 | |
| BFLWXPJTAKXXKT-UHFFFAOYSA-N | |
| 184268 | |
| COC1=CC=CC(=C1N)N |
| C7H10N2O | |
| MFCD08276903 | |
| 2,3-diaminoanisole, 2,3-diaminoanisol, diaminoanisole, 3-methoxy-benzene-1,2-diamine, 3-methoxy-1,2-phenylenediamine, 1,2-benzenediamine, 3-methoxy, 3-methoxy-o-phenylenediamine, 3-methoxy-1,2-benzenediamine, ambkkkkk251, 2-amino-3-methoxyaniline | |
| 3-methoxybenzene-1,2-diamine |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only