missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-o-phenylenediamine, 97%

Catalog No. AAH35286MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH35286MD 250 mg
AAH3528603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH35286MD Supplier Thermo Scientific Chemicals Supplier No. H35286MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 37466-89-0 | C7H10N2O | 138.17 g/mol

3-Methoxy-o-phenylenediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-o-phenylenediamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents.

Chemical Identifiers

CAS 37466-89-0
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD08276903
InChI Key BFLWXPJTAKXXKT-UHFFFAOYSA-N
Synonym 2,3-diaminoanisole, 2,3-diaminoanisol, diaminoanisole, 3-methoxy-benzene-1,2-diamine, 3-methoxy-1,2-phenylenediamine, 1,2-benzenediamine, 3-methoxy, 3-methoxy-o-phenylenediamine, 3-methoxy-1,2-benzenediamine, ambkkkkk251, 2-amino-3-methoxyaniline
PubChem CID 184268
IUPAC Name 3-methoxybenzene-1,2-diamine
SMILES COC1=CC=CC(=C1N)N

Specifications

Melting Point 39°C to 40°C
Quantity 250 mg
UN Number UN2811
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 3-Methoxy-o-phenylenediamine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only