missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-4-methylbenzoic Acid 98.0+%, TCI America™

Supplier:  TCI America M14265G

 View more versions of this product

Catalog No. M14265G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Methoxy-4-methylbenzoic Acid
White
MFCD00002566
4-methyl-m-anisic acid, 3-methoxy-p-toluic acid, benzoic acid, 3-methoxy-4-methyl, 3-methoxy-4-methyl-benzoic acid, 3-methoxy-4-methyl benzoic acid, 3-methoxy-p-toluic acid; 4-methyl-m-anisic acid, 3-methoxy-4-methylbenzoicacid, pubchem2672, 5-carboxy-2-methylanisole, acmc-209oj0
COC1=CC(=CC=C1C)C(O)=O
166.18
166.18
Crystalline Powder
7151-68-0
C9H10O3
5 g
CEAVPXDEPGAVDA-UHFFFAOYSA-N
3-methoxy-4-methylbenzoic acid
81579
≥98.0% (HPLC,T)

Chemical Identifiers

7151-68-0
166.18
CEAVPXDEPGAVDA-UHFFFAOYSA-N
81579
COC1=CC(=CC=C1C)C(O)=O
C9H10O3
MFCD00002566
4-methyl-m-anisic acid, 3-methoxy-p-toluic acid, benzoic acid, 3-methoxy-4-methyl, 3-methoxy-4-methyl-benzoic acid, 3-methoxy-4-methyl benzoic acid, 3-methoxy-p-toluic acid; 4-methyl-m-anisic acid, 3-methoxy-4-methylbenzoicacid, pubchem2672, 5-carboxy-2-methylanisole, acmc-209oj0
3-methoxy-4-methylbenzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions