missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-4-methylbenzoic acid, 99%

Catalog No. p-7026772
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2298206 5 g
AAB2298214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2298206 Supplier Thermo Scientific Chemicals Supplier No. B2298206

CAS: 7151-68-0 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7151-68-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00002566
InChI Key CEAVPXDEPGAVDA-UHFFFAOYSA-N
Synonym 4-methyl-m-anisic acid, 3-methoxy-p-toluic acid, benzoic acid, 3-methoxy-4-methyl, 3-methoxy-4-methyl-benzoic acid, 3-methoxy-4-methyl benzoic acid, 3-methoxy-p-toluic acid; 4-methyl-m-anisic acid, 3-methoxy-4-methylbenzoicacid, pubchem2672, 5-carboxy-2-methylanisole, acmc-209oj0
PubChem CID 81579
IUPAC Name 3-methoxy-4-methylbenzoic acid
SMILES COC1=CC(=CC=C1C)C(O)=O

Specifications

Melting Point 158°C to 160°C
Quantity 5 g
Beilstein 1938389
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material 3-Methoxy-4-methylbenzoic acid
EINECSNumber 230-486-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only