missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-3-methylbutyl Acetate 98.0+%, TCI America™

Catalog No. p-7117159
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A096425ML 25 mL
A0964500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A096425ML Supplier TCI America Supplier No. A096425ML

Chemical Identifiers

CAS 103429-90-9
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.213
MDL Number MFCD00059340
InChI Key RYNQKSJRFHJZTK-UHFFFAOYSA-N
Synonym Acetic Acid 3-Methoxy-3-methylbutyl Ester
PubChem CID 62308
IUPAC Name (3-methoxy-3-methylbutyl) acetate
SMILES CC(=O)OCCC(C)(C)OC

Specifications

Color Colorless
Boiling Point 74°C
Quantity 25 mL
Formula Weight 160.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxy-3-methylbutyl Acetate
EINECSNumber (2)-3291
TSCA No