accessibility menu, dialog, popup

UserName

CH3OH3CCH3OCH3O

CAS RN 103429-90-9

IUPAC Name: 3-methoxy-3-methylbutyl acetate
Synonyms: 3-methoxy-3-methylbutyl acetate
Molecular Weight (g/mol): 160.21
Molecular Formula: C8H16O3
InChi Key: RYNQKSJRFHJZTK-UHFFFAOYSA-N
SMILES: COC(C)(C)CCOC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results