missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-3-methylbutanol 98.0+%, TCI America™

Catalog No. p-7132304
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
M069525ML 25 mL
M0695500ML 500 mL
2 options

Catalog No. M069525ML

Supplier: TCI America M069525ML

Only null left

Chemical Identifiers

CAS 56539-66-3
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00044771
InChI Key MFKRHJVUCZRDTF-UHFFFAOYSA-N
Synonym 3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389
PubChem CID 62118
IUPAC Name 3-methoxy-3-methylbutan-1-ol
SMILES CC(C)(CCO)OC

Specifications

Color Colorless
Boiling Point 174°C
Quantity 25 mL
Formula Weight 118.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxy-3-methylbutanol
EINECSNumber (2)-3079
TSCA Yes