accessibility menu, dialog, popup

UserName

CAS RN 56539-66-3

IUPAC Name: 3-methoxy-3-methylbutan-1-ol
Synonyms: methoxymethylbutanol
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: MFKRHJVUCZRDTF-UHFFFAOYSA-N
SMILES: COC(C)(C)CCO
Boiling Point 
  • (3)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)