missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-3-methylbutanol 98.0+%, TCI America™

Catalog No. M069525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M069525ML 25 mL
M0695500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M069525ML Supplier TCI America Supplier No. M069525ML

Chemical Identifiers

CAS 56539-66-3
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00044771
InChI Key MFKRHJVUCZRDTF-UHFFFAOYSA-N
Synonym 3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389
PubChem CID 62118
IUPAC Name 3-methoxy-3-methylbutan-1-ol
SMILES CC(C)(CCO)OC

Specifications

Color Colorless
Boiling Point 174°C
Quantity 25 mL
Formula Weight 118.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxy-3-methylbutanol
EINECSNumber (2)-3079
TSCA Yes