missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-3-methyl-1-butanol, 98+%

Catalog No. AAL15953AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15953AP 500 mL
AAL15953AE 100 mL
AAL159530F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL15953AP Supplier Thermo Scientific Chemicals Supplier No. L15953AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56539-66-3 | C6H14O2 | 118.176 g/mol

3-Methoxy-3-methyl-1-butanol is a solvent that compares favorably with its isomer ethylene glycol monobutyl ether. It is employed as raw material for the production of industrial detergents. It is also used as solvent for paints, inks, and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-3-methyl-1-butanol is a solvent that compares favorably with its isomer ethylene glycol monobutyl ether. It is employed as raw material for the production of industrial detergents. It is also used as solvent for paints, inks, and fragrances.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Chemical Identifiers

CAS 56539-66-3
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00044771
InChI Key MFKRHJVUCZRDTF-UHFFFAOYSA-N
Synonym 3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389
PubChem CID 62118
IUPAC Name 3-methoxy-3-methylbutan-1-ol
SMILES CC(C)(CCO)OC

Specifications

Density 0.926
Boiling Point 173°C to 175°C
Flash Point 71°C (159°F)
Refractive Index 1.428
Quantity 500 mL
Beilstein 1732882
Solubility Information Soluble in water.
Formula Weight 118.18
Percent Purity ≥98%
Chemical Name or Material 3-Methoxy-3-methyl-1-butanol
Hazard Category H227-H319
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
EINECSNumber 260-252-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only