missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate 95.0+%, TCI America™

Catalog No. p-7132892
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
M18841G 1 g
M18845G 5 g
2 options

Catalog No. M18841G

Supplier: TCI America M18841G

Only null left

Chemical Identifiers

CAS 217813-03-1
Molecular Formula C11H15F3O4SSi
Molecular Weight (g/mol) 328.38
MDL Number MFCD10566916
InChI Key OJVREKSBMMQLBJ-UHFFFAOYSA-N
Synonym 3-Methoxy-2-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 3-Methoxy-2-(trimethylsilyl)phenyl Ester
PubChem CID 11023933
IUPAC Name 3-methoxy-2-(trimethylsilyl)phenyl trifluoromethanesulfonate
SMILES COC1=C(C(OS(=O)(=O)C(F)(F)F)=CC=C1)[Si](C)(C)C

Specifications

Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 328.38
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxy-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate
TSCA No
Recommended Storage Refrigerator