CAS RN 217813-03-1
IUPAC Name:
3-methoxy-2-(trimethylsilyl)phenyl trifluoromethanesulfonate
Synonyms:
3-methoxy-2-(trimethylsilyl)phenyl trifluoromethanesulfonate
Molecular Weight (g/mol):
328.38
Molecular Formula:
C11H15F3O4SSi
InChi Key:
OJVREKSBMMQLBJ-UHFFFAOYSA-N
SMILES:
COC1=C(C(OS(=O)(=O)C(F)(F)F)=CC=C1)[Si](C)(C)C
1
–
1
of
1
results
1
3-Methoxy-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate 95.0+%, TCI America™
CAS: 217813-03-1 Molecular Formula: C11H15F3O4SSi Molecular Weight (g/mol): 328.38 MDL Number: MFCD10566916 InChI Key: OJVREKSBMMQLBJ-UHFFFAOYSA-N Synonym: 3-Methoxy-2-(trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 3-Methoxy-2-(trimethylsilyl)phenyl Ester PubChem CID: 11023933 IUPAC Name: 3-methoxy-2-(trimethylsilyl)phenyl trifluoromethanesulfonate SMILES: COC1=C(C(OS(=O)(=O)C(F)(F)F)=CC=C1)[Si](C)(C)C