missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxy-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| UN Number | 3265 |
| Formula Weight | 328.38 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Methoxy-2-(trimethylsilyl)phenyl Trifluoromethanesulfonate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |