missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-2-naphthoic Acid 98.0+%, TCI America™

Catalog No. M14615G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14615G 5 g
M14611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14615G Supplier TCI America Supplier No. M14615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 883-62-5
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.21
MDL Number MFCD00094168
InChI Key RTBQQRFTCVDODF-UHFFFAOYSA-N
PubChem CID 70161
IUPAC Name 3-methoxynaphthalene-2-carboxylic acid
SMILES COC1=C(C=C2C=CC=CC2=C1)C(O)=O

Specifications

Melting Point 134°C
Color Yellow
Quantity 5 g
Formula Weight 202.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxy-2-naphthoic Acid