missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxy-1-propanol 98.0+%, TCI America™
$84.70 - $242.57
Chemical Identifiers
| CAS | 1589-49-7 |
|---|---|
| Molecular Formula | C4H10O2 |
| Molecular Weight (g/mol) | 90.122 |
| MDL Number | MFCD00039550 |
| InChI Key | JDFDHBSESGTDAL-UHFFFAOYSA-N |
| Synonym | 3-methoxy-1-propanol, 1-propanol, 3-methoxy, 3-methoxypropanol, trimethylene glycol monomethyl ether, beta-pgme, 1,3-propanediol monomethyl ether, unii-oak281wz1p, propylene glycol monomethyl ether, beta, oak281wz1p, mpol |
| PubChem CID | 74116 |
| IUPAC Name | 3-methoxypropan-1-ol |
| SMILES | COCCCO |
Chemical Identifiers
| 1589-49-7 | |
| 90.122 | |
| JDFDHBSESGTDAL-UHFFFAOYSA-N | |
| 74116 | |
| COCCCO |
| C4H10O2 | |
| MFCD00039550 | |
| 3-methoxy-1-propanol, 1-propanol, 3-methoxy, 3-methoxypropanol, trimethylene glycol monomethyl ether, beta-pgme, 1,3-propanediol monomethyl ether, unii-oak281wz1p, propylene glycol monomethyl ether, beta, oak281wz1p, mpol | |
| 3-methoxypropan-1-ol |
Specifications
| 1589-49-7 | |
| 153°C | |
| MFCD00039550 | |
| 1993 | |
| JDFDHBSESGTDAL-UHFFFAOYSA-N | |
| 3-methoxypropan-1-ol | |
| 74116 | |
| ≥98.0% (GC) | |
| 3-Methoxy-1-propanol |
| Yellow | |
| C4H10O2 | |
| 25 g | |
| 3-methoxy-1-propanol, 1-propanol, 3-methoxy, 3-methoxypropanol, trimethylene glycol monomethyl ether, beta-pgme, 1,3-propanediol monomethyl ether, unii-oak281wz1p, propylene glycol monomethyl ether, beta, oak281wz1p, mpol | |
| COCCCO | |
| 90.122 | |
| 90.12 | |
| Liquid |
Safety and Handling
RTECSNumber : UB7650000
TSCA : No