accessibility menu, dialog, popup

UserName

H3COOH

CAS RN 1589-49-7

IUPAC Name: 3-methoxypropan-1-ol
Synonyms: 1-propanol, 3-methoxy-
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: JDFDHBSESGTDAL-UHFFFAOYSA-N
SMILES: COCCCO
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)