missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-1-propanol 98.0+%, TCI America™

Catalog No. M147525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M147525G 25 g
M1475250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M147525G Supplier TCI America Supplier No. M147525G

Chemical Identifiers

CAS 1589-49-7
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00039550
InChI Key JDFDHBSESGTDAL-UHFFFAOYSA-N
Synonym 3-methoxy-1-propanol, 1-propanol, 3-methoxy, 3-methoxypropanol, trimethylene glycol monomethyl ether, beta-pgme, 1,3-propanediol monomethyl ether, unii-oak281wz1p, propylene glycol monomethyl ether, beta, oak281wz1p, mpol
PubChem CID 74116
IUPAC Name 3-methoxypropan-1-ol
SMILES COCCCO

Specifications

Color Yellow
Boiling Point 153°C
Quantity 25 g
UN Number 1993
Formula Weight 90.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxy-1-propanol
RTECSNumber UB7650000
TSCA No