missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-1-butanol 98.0+%, TCI America™

Catalog No. M0109500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0109500ML 500 mL
M010925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0109500ML Supplier TCI America Supplier No. M0109500ML

Chemical Identifiers

CAS 2517-43-3
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00002931
InChI Key JSGVZVOGOQILFM-UHFFFAOYNA-N
Synonym 3-methoxy-1-butanol, 3-methoxybutanol, 1-butanol, 3-methoxy, methoxybutanol, ccris 8976, 1-butanol, 3-methoxy-,-, 1-butanol, 3-methoxy-, r, 3-methoxy-butanol, 3-methoxybutane-1-ol, 3-methoxybutyl alcohol
PubChem CID 17291
IUPAC Name 3-methoxybutan-1-ol
SMILES COC(C)CCO

Specifications

Color Colorless
Boiling Point 158°C
Quantity 500 mL
Formula Weight 104.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxy-1-butanol
EINECSNumber (2)-0409
TSCA Yes