missing translation for 'onlineSavingsMsg'
Learn More

(3-Mercaptopropyl)trimethoxysilane 96.0+%, TCI America™

Supplier:  TCI America M0928100ML

 View more versions of this product

Catalog No. M0928100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(3-Mercaptopropyl)trimethoxysilane
Colorless
C6H16O3SSi
100 mL
trimethoxysilylpropanethiol, 3-mercaptopropyltrimethoxysilane, 3-mercaptopropyl trimethoxysilane, 1-propanethiol, 3-trimethoxysilyl, silquest a 189, 3-trimethoxysilyl propanethiol, prosil 196, silane a 189, union carbide a-189, sila-ace s 810
CO[Si](CCCS)(OC)OC
196.336
196.34
Liquid
4420-74-0
215°C
MFCD00004901
3082
UUEWCQRISZBELL-UHFFFAOYSA-N
3-trimethoxysilylpropane-1-thiol
20473
≥96.0% (GC)

Chemical Identifiers

4420-74-0
196.336
UUEWCQRISZBELL-UHFFFAOYSA-N
20473
CO[Si](CCCS)(OC)OC
C6H16O3SSi
MFCD00004901
trimethoxysilylpropanethiol, 3-mercaptopropyltrimethoxysilane, 3-mercaptopropyl trimethoxysilane, 1-propanethiol, 3-trimethoxysilyl, silquest a 189, 3-trimethoxysilyl propanethiol, prosil 196, silane a 189, union carbide a-189, sila-ace s 810
3-trimethoxysilylpropane-1-thiol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2045

RTECSNumber : TZ7800000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions