missing translation for 'onlineSavingsMsg'
Learn More

(3-Mercaptopropyl)trimethoxysilane 96.0+%, TCI America™

Catalog No. p-7132446
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M092825ML 25 mL
M0928100ML 100 mL
M0928500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M092825ML Supplier TCI America Supplier No. M092825ML
Only null left

Chemical Identifiers

CAS 4420-74-0
Molecular Formula C6H16O3SSi
Molecular Weight (g/mol) 196.336
MDL Number MFCD00004901
InChI Key UUEWCQRISZBELL-UHFFFAOYSA-N
Synonym trimethoxysilylpropanethiol, 3-mercaptopropyltrimethoxysilane, 3-mercaptopropyl trimethoxysilane, 1-propanethiol, 3-trimethoxysilyl, silquest a 189, 3-trimethoxysilyl propanethiol, prosil 196, silane a 189, union carbide a-189, sila-ace s 810
PubChem CID 20473
IUPAC Name 3-trimethoxysilylpropane-1-thiol
SMILES CO[Si](CCCS)(OC)OC

Specifications

Color Colorless
Boiling Point 215°C
Quantity 25 mL
UN Number 3082
Formula Weight 196.34
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (3-Mercaptopropyl)trimethoxysilane
EINECSNumber (2)-2045
RTECSNumber TZ7800000
TSCA Yes