missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Mercapto-2-pentanone 97.0+%, TCI America™
Supplier: TCI America M20265G
Specifications
| 3-Mercapto-2-pentanone | |
| Yellow | |
| C5H10OS | |
| 5 g | |
| 3-mercapto-2-pentanone, 2-pentanone, 3-mercapto, 3-mercaptopentan-2-one, fema no. 3300, 3-mercapto-2-penanone, 2-pentanone,3-mercapto, acmc-1b6ys, 2-pentanone, 3-mercapto-7ci,9ci | |
| CCC(C(=O)C)S | |
| 118.194 | |
| 118.19 | |
| Liquid |
| 67633-97-0 | |
| 52°C | |
| MFCD02093290 | |
| 3336 | |
| SZECUQRKLXRGSJ-UHFFFAOYSA-N | |
| 3-sulfanylpentan-2-one | |
| 62237 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 67633-97-0 | |
| 118.194 | |
| SZECUQRKLXRGSJ-UHFFFAOYSA-N | |
| 62237 | |
| CCC(C(=O)C)S |
| C5H10OS | |
| MFCD02093290 | |
| 3-mercapto-2-pentanone, 2-pentanone, 3-mercapto, 3-mercaptopentan-2-one, fema no. 3300, 3-mercapto-2-penanone, 2-pentanone,3-mercapto, acmc-1b6ys, 2-pentanone, 3-mercapto-7ci,9ci | |
| 3-sulfanylpentan-2-one |
Safety and Handling
TSCA : Yes