missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1-propanol 97.0+%, TCI America™

Catalog No. p-7132599
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
M12065G 5 g
M120625G 25 g
2 options

Catalog No. M12065G

Supplier: TCI America M12065G

Only null left

Chemical Identifiers

CAS 19721-22-3
Molecular Formula C3H8OS
Molecular Weight (g/mol) 92.156
MDL Number MFCD00192262
InChI Key SHLSSLVZXJBVHE-UHFFFAOYSA-N
Synonym 3-mercapto-1-propanol, 1-propanol, 3-mercapto, 3-mercaptopropanol, 3-hydroxypropanethiol, 3-hydroxypropyl mercaptan, 3-mecapto-1-propanol, 3-mecaptopropan-1-ol, 1-mercapto-3-propanol, 3-hydroxypropylmercaptan, 3-mercaptopropan-1-ol
PubChem CID 88211
IUPAC Name 3-sulfanylpropan-1-ol
SMILES C(CO)CS

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
UN Number 2810
Formula Weight 92.16
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Mercapto-1-propanol
RTECSNumber UB7630000
TSCA Yes