missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1-propanol, 97+%

Catalog No. ac388570010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC388570010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC388570010 Supplier Thermo Scientific Chemicals Supplier No. 388570010
Only null left

CAS: 19721-22-3 | C3H8OS | 92.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19721-22-3
Molecular Formula C3H8OS
Molecular Weight (g/mol) 92.16
MDL Number MFCD00192262
InChI Key SHLSSLVZXJBVHE-UHFFFAOYSA-N
Synonym 3-mercapto-1-propanol, 1-propanol, 3-mercapto, 3-mercaptopropanol, 3-hydroxypropanethiol, 3-hydroxypropyl mercaptan, 3-mecapto-1-propanol, 3-mecaptopropan-1-ol, 1-mercapto-3-propanol, 3-hydroxypropylmercaptan, 3-mercaptopropan-1-ol
PubChem CID 88211
IUPAC Name 3-sulfanylpropan-1-ol
SMILES C(CO)CS

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless
Density 1.0620g/mL
Boiling Point 106°C (50.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4950 to 1.497
Linear Formula SH(CH2)3OH
Quantity 1 g
Specific Gravity 1.062
Solubility Information Solubility in water: slightly soluble
Formula Weight 92.16
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material 3-Mercapto-1-propanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Toxic in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only