missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Maleimidopropionic Acid 97.0+%, TCI America™
$46.20 - $525.57
Chemical Identifiers
| CAS | 7423-55-4 |
|---|---|
| Molecular Formula | C7H7NO4 |
| Molecular Weight (g/mol) | 169.14 |
| MDL Number | MFCD00043030 |
| InChI Key | IUTPJBLLJJNPAJ-UHFFFAOYSA-N |
| Synonym | 3-maleimidopropionic acid, 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoic acid, n-maleoyl-beta-alanine, 3-2,5-dioxopyrrol-1-yl propanoic acid, 1h-pyrrole-1-propanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 2-cooh, n-carboxyethylmaleimide, 3-maleimdepropionicacid, pubchem11845, acmc-209otw |
| PubChem CID | 573621 |
| IUPAC Name | 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoic acid |
| SMILES | OC(=O)CCN1C(=O)C=CC1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M1962200MG
|
TCI America
M1962200MG |
200 mg |
Each for $46.20
|
|
|||||
|
M19621G
|
TCI America
M19621G |
1 g |
Each for $154.02
|
|
|||||
|
M19625G
|
TCI America
M19625G |
5 g |
Each for $525.57
|
|
|||||
Chemical Identifiers
| 7423-55-4 | |
| 169.14 | |
| IUTPJBLLJJNPAJ-UHFFFAOYSA-N | |
| 573621 | |
| OC(=O)CCN1C(=O)C=CC1=O |
| C7H7NO4 | |
| MFCD00043030 | |
| 3-maleimidopropionic acid, 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoic acid, n-maleoyl-beta-alanine, 3-2,5-dioxopyrrol-1-yl propanoic acid, 1h-pyrrole-1-propanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 2-cooh, n-carboxyethylmaleimide, 3-maleimdepropionicacid, pubchem11845, acmc-209otw | |
| 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoic acid |
Specifications
| 7423-55-4 | |
| White | |
| MFCD00043030 | |
| 3-maleimidopropionic acid, 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoic acid, n-maleoyl-beta-alanine, 3-2,5-dioxopyrrol-1-yl propanoic acid, 1h-pyrrole-1-propanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 2-cooh, n-carboxyethylmaleimide, 3-maleimdepropionicacid, pubchem11845, acmc-209otw | |
| OC(=O)CCN1C(=O)C=CC1=O | |
| 169.14 | |
| 169.14 | |
| Crystalline Powder |
| 110°C | |
| C7H7NO4 | |
| 200 mg | |
| IUTPJBLLJJNPAJ-UHFFFAOYSA-N | |
| 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoic acid | |
| 573621 | |
| ≥97.0% (GC) | |
| 3-Maleimidopropionic Acid |
Safety and Handling
TSCA : No