Learn More
3-Maleimidopropionic acid, 95%
CAS: 7423-55-4 | C7H7NO4 | 169.14 g/mol
$310.70 - $1,267.49
Chemical Identifiers
| CAS | 7423-55-4 |
|---|---|
| Molecular Formula | C7H7NO4 |
| Molecular Weight (g/mol) | 169.14 |
| MDL Number | MFCD00043030 |
| InChI Key | IUTPJBLLJJNPAJ-UHFFFAOYSA-N |
| Synonym | 3-maleimidopropionic acid, 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoic acid, n-maleoyl-beta-alanine, 3-2,5-dioxopyrrol-1-yl propanoic acid, 1h-pyrrole-1-propanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 2-cooh, n-carboxyethylmaleimide, 3-maleimdepropionicacid, pubchem11845, acmc-209otw |
| PubChem CID | 573621 |
| IUPAC Name | 3-(2,5-dioxopyrrol-1-yl)propanoic acid |
| SMILES | OC(=O)CCN1C(=O)C=CC1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6153603
|
Thermo Scientific Chemicals
H6153603 |
1 g |
N/A
|
|
|||||
|
AAH6153606
|
Thermo Scientific Chemicals
H6153606 |
5 g |
N/A
|
|
|||||
Description
3-Maleimidopropionic acid is a sulfhydryl reactive heterobifunctional crosslinking reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Maleimidopropionic acid is a sulfhydryl reactive heterobifunctional crosslinking reagent.
Solubility
Soluble in methanol, dimethyl formamide, alcohol and dimethyl sulfoxide. Slightly soluble in water.
Notes
Moisture sensitive. Store in cool place.
Chemical Identifiers
| 7423-55-4 | |
| 169.14 | |
| IUTPJBLLJJNPAJ-UHFFFAOYSA-N | |
| 573621 | |
| OC(=O)CCN1C(=O)C=CC1=O |
| C7H7NO4 | |
| MFCD00043030 | |
| 3-maleimidopropionic acid, 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoic acid, n-maleoyl-beta-alanine, 3-2,5-dioxopyrrol-1-yl propanoic acid, 1h-pyrrole-1-propanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 2-cooh, n-carboxyethylmaleimide, 3-maleimdepropionicacid, pubchem11845, acmc-209otw | |
| 3-(2,5-dioxopyrrol-1-yl)propanoic acid |
Specifications
| 7423-55-4 | |
| White | |
| MFCD00043030 | |
| 1528952 | |
| 3-maleimidopropionic acid, 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoic acid, n-maleoyl-beta-alanine, 3-2,5-dioxopyrrol-1-yl propanoic acid, 1h-pyrrole-1-propanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 2-cooh, n-carboxyethylmaleimide, 3-maleimdepropionicacid, pubchem11845, acmc-209otw | |
| IUTPJBLLJJNPAJ-UHFFFAOYSA-N | |
| 3-(2,5-dioxopyrrol-1-yl)propanoic acid | |
| 573621 | |
| 95% | |
| 3-Maleimidopropionic acid |
| 108°C to 112°C | |
| C7H7NO4 | |
| 1 g | |
| Moisture sensitive | |
| Soluble in methanol,dimethyl formamide,alcohol and dimethyl sulfoxide. Slightly soluble in water. | |
| OC(=O)CCN1C(=O)C=CC1=O | |
| 169.14 | |
| 169.13 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only