Learn More
3-Isopropylbenzeneboronic acid, 99%
CAS: 216019-28-2 | C9H13BO2 | 164.01 g/mol
Supplier: Thermo Scientific Chemicals L15530MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Isopropylbenzeneboronic acid | |
| ∼75°C to 80°C | |
| C9H13BO2 | |
| 250 mg | |
| 3-isopropylphenylboronic acid, 3-isopropylbenzeneboronic acid, 3-cumylboronic acid, 3-isopropylphenyl boronic acid, 3-isopropylphebylboronic acid, 3-propan-2-yl phenyl boranediol, boronic acid, 3-1-methylethyl phenyl, boronic acid, b-3-propylphenyl, 3-propan-2-yl phenyl boronic acid, 3-propan-2-ylphenyl boronic acid | |
| CC(C)C1=CC=CC(=C1)B(O)O | |
| 164.01 | |
| 164.01 |
| 216019-28-2 | |
| >110°C (230°F) | |
| MFCD01074634 | |
| 3090001 | |
| QSWLFBMVIGQONC-UHFFFAOYSA-N | |
| (3-propan-2-ylphenyl)boronic acid | |
| 2773478 | |
| 99% |
Chemical Identifiers
| 216019-28-2 | |
| 164.01 | |
| QSWLFBMVIGQONC-UHFFFAOYSA-N | |
| 2773478 | |
| CC(C)C1=CC=CC(=C1)B(O)O |
| C9H13BO2 | |
| MFCD01074634 | |
| 3-isopropylphenylboronic acid, 3-isopropylbenzeneboronic acid, 3-cumylboronic acid, 3-isopropylphenyl boronic acid, 3-isopropylphebylboronic acid, 3-propan-2-yl phenyl boranediol, boronic acid, 3-1-methylethyl phenyl, boronic acid, b-3-propylphenyl, 3-propan-2-yl phenyl boronic acid, 3-propan-2-ylphenyl boronic acid | |
| (3-propan-2-ylphenyl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only