missing translation for 'onlineSavingsMsg'
Learn More

3-Isopropylaniline 98.0+%, TCI America™

Catalog No. p-7131404
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I043325ML 25 mL
I0433500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I043325ML Supplier TCI America Supplier No. I043325ML
Only null left

Chemical Identifiers

CAS 5369-16-4
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00092918
InChI Key XCCNRBCNYGWTQX-UHFFFAOYSA-N
Synonym 3-Aminocumene, m-Cumidine
PubChem CID 458679
IUPAC Name 3-(propan-2-yl)aniline
SMILES CC(C)C1=CC(N)=CC=C1

Specifications

Color Yellow
Boiling Point 80°C
Quantity 25 mL
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Isopropylaniline
EINECSNumber (3)-0206
TSCA No