missing translation for 'onlineSavingsMsg'
Learn More

3-Isopropylaniline 98.0+%, TCI America™

Numéro de catalogue. I0433500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
I0433500ML 500 mL
I043325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. I0433500ML Fournisseur TCI America Code fournisseur I0433500ML
Il en reste null

Identifiants chimiques

CAS 5369-16-4
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00092918
InChI Key XCCNRBCNYGWTQX-UHFFFAOYSA-N
Synonym 3-Aminocumene, m-Cumidine
PubChem CID 458679
IUPAC Name 3-(propan-2-yl)aniline
SMILES CC(C)C1=CC(N)=CC=C1

Spécifications

Color Yellow
Boiling Point 80°C
Quantity 500 mL
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Isopropylaniline
EINECSNumber (3)-0206
TSCA No