missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Isopropylaniline 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 80°C |
| Quantity | 25 mL |
| Formula Weight | 135.21 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Isopropylaniline |
Safety and Handling
| EINECSNumber | (3)-0206 |
| TSCA | No |