missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Isopropoxypropylamine 98.0+%, TCI America™
$57.79 - $158.44
Chemical Identifiers
| CAS | 2906-12-9 |
|---|---|
| Molecular Formula | C6H15NO |
| Molecular Weight (g/mol) | 117.19 |
| MDL Number | MFCD00008220 |
| InChI Key | VHYUNSUGCNKWSO-UHFFFAOYSA-N |
| Synonym | 3-isopropoxypropylamine, 3-isopropoxy-1-propylamine, isopropoxypropylamine, 1-propanamine, 3-1-methylethoxy, 3-aminopropyl isopropyl ether, 3-1-methylethoxy-1-propanamine, 3-isopropoxy-n-propylamine, 3-isopropoxypropan-1-amine, propylamine, 3-isopropoxy, 3-propan-2-yloxy propan-1-amine |
| PubChem CID | 76194 |
| IUPAC Name | 3-propan-2-yloxypropan-1-amine |
| SMILES | CC(C)OCCCN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I016225ML
|
TCI America
I016225ML |
25 mL |
Each for $57.79
|
|
|||||
|
I0162500ML
|
TCI America
I0162500ML |
500 mL |
Each for $158.44
|
|
|||||
Chemical Identifiers
| 2906-12-9 | |
| 117.19 | |
| VHYUNSUGCNKWSO-UHFFFAOYSA-N | |
| 76194 | |
| CC(C)OCCCN |
| C6H15NO | |
| MFCD00008220 | |
| 3-isopropoxypropylamine, 3-isopropoxy-1-propylamine, isopropoxypropylamine, 1-propanamine, 3-1-methylethoxy, 3-aminopropyl isopropyl ether, 3-1-methylethoxy-1-propanamine, 3-isopropoxy-n-propylamine, 3-isopropoxypropan-1-amine, propylamine, 3-isopropoxy, 3-propan-2-yloxy propan-1-amine | |
| 3-propan-2-yloxypropan-1-amine |
Specifications
| 2906-12-9 | |
| 144°C | |
| MFCD00008220 | |
| 2734 | |
| VHYUNSUGCNKWSO-UHFFFAOYSA-N | |
| 3-propan-2-yloxypropan-1-amine | |
| 76194 | |
| ≥98.0% (GC,T) | |
| 3-Isopropoxypropylamine |
| Yellow | |
| C6H15NO | |
| 25 mL | |
| 3-isopropoxypropylamine, 3-isopropoxy-1-propylamine, isopropoxypropylamine, 1-propanamine, 3-1-methylethoxy, 3-aminopropyl isopropyl ether, 3-1-methylethoxy-1-propanamine, 3-isopropoxy-n-propylamine, 3-isopropoxypropan-1-amine, propylamine, 3-isopropoxy, 3-propan-2-yloxy propan-1-amine | |
| CC(C)OCCCN | |
| 117.19 | |
| 117.19 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0385
TSCA : Yes