accessibility menu, dialog, popup

UserName

CAS RN 2906-12-9

IUPAC Name: 3-(propan-2-yloxy)propan-1-amine
Synonyms: 3-isopropoxypropan-1-amine
Molecular Weight (g/mol): 117.19
Molecular Formula: C6H15NO
InChi Key: VHYUNSUGCNKWSO-UHFFFAOYSA-N
SMILES: CC(C)OCCCN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results