missing translation for 'onlineSavingsMsg'
Learn More

3-Isopropoxypropylamine 98.0+%, TCI America™

Catalog No. I016225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I016225ML 25 mL
I0162500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I016225ML Supplier TCI America Supplier No. I016225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2906-12-9
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
MDL Number MFCD00008220
InChI Key VHYUNSUGCNKWSO-UHFFFAOYSA-N
Synonym 3-isopropoxypropylamine, 3-isopropoxy-1-propylamine, isopropoxypropylamine, 1-propanamine, 3-1-methylethoxy, 3-aminopropyl isopropyl ether, 3-1-methylethoxy-1-propanamine, 3-isopropoxy-n-propylamine, 3-isopropoxypropan-1-amine, propylamine, 3-isopropoxy, 3-propan-2-yloxy propan-1-amine
PubChem CID 76194
IUPAC Name 3-propan-2-yloxypropan-1-amine
SMILES CC(C)OCCCN

Specifications

Color Yellow
Boiling Point 144°C
Quantity 25 mL
UN Number 2734
Formula Weight 117.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Isopropoxypropylamine
EINECSNumber (2)-0385
TSCA Yes