Learn More
(3-Isocyanatopropyl)triethoxysilane, 95%
CAS: 24801-88-5 | C10H21NO4Si | 247.366 g/mol
Supplier: Thermo Scientific Chemicals A1289006
Description
It finds its application as a component in hybrid organic/inorganic urethanes. It is also used in the synthesis of a polymer surface containing covalently attached triethoxysilane functionality: adhesion to glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application as a component in hybrid organic/inorganic urethanes. It is also used in the synthesis of a polymer surface containing covalently attached triethoxysilane functionality: adhesion to glass.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from alcohols, amines, oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
| 238°C | |
| 24801-88-5 | |
| C10H21NO4Si | |
| MFCD00051459 | |
| 4404652 | |
| 3-isocyanatopropyltriethoxysilane, triethoxy 3-isocyanatopropyl silane, 3-isocyanatopropyl triethoxysilane, isocyanatopropyltriethoxysilane, 3-triethoxysilyl propyl isocyanate, silane, triethoxy 3-isocyanatopropyl, unii-9br6002p6e, isocyanic acid, 3-triethoxysilyl propylester, isocyanic acid, 3-triethoxysilyl propyl ester, icptes | |
| FRGPKMWIYVTFIQ-UHFFFAOYSA-N | |
| triethoxy(3-isocyanatopropyl)silane | |
| 90613 | |
| 95% | |
| 1.421 | |
| 0.99 |
| (3-Isocyanatopropyl)triethoxysilane | |
| 5 g | |
| (CH3CH2O)3SiCH2CH2CH2NCO | |
| UN2927 | |
| 77°C (170°F) | |
| Hydrolyzes in water. | |
| CCO[Si](CCCN=C=O)(OCC)OCC | |
| 247.366 | |
| 247.37 | |
| Mild | |
| Moisture sensitive |
Chemical Identifiers
| 24801-88-5 | |
| 247.366 | |
| FRGPKMWIYVTFIQ-UHFFFAOYSA-N | |
| 90613 | |
| CCO[Si](CCCN=C=O)(OCC)OCC |
| C10H21NO4Si | |
| MFCD00051459 | |
| 3-isocyanatopropyltriethoxysilane, triethoxy 3-isocyanatopropyl silane, 3-isocyanatopropyl triethoxysilane, isocyanatopropyltriethoxysilane, 3-triethoxysilyl propyl isocyanate, silane, triethoxy 3-isocyanatopropyl, unii-9br6002p6e, isocyanic acid, 3-triethoxysilyl propylester, isocyanic acid, 3-triethoxysilyl propyl ester, icptes | |
| triethoxy(3-isocyanatopropyl)silane |
Safety and Handling
GHS H Statement
H330-H334-H314-H318-H227-H302-H312
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
P210-P235-P260-P264b-P270-P271-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312-H314-H330-H334
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber : 246-467-6
RTECSNumber : VV6691000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only