accessibility menu, dialog, popup

UserName

CH3OSiNCOOCH3OH3C

CAS RN 24801-88-5

IUPAC Name: triethoxy(3-isocyanatopropyl)silane
Synonyms: triethoxy(3-isocyanatopropyl)silane
Molecular Weight (g/mol): 247.37
Molecular Formula: C10H21NO4Si
InChi Key: FRGPKMWIYVTFIQ-UHFFFAOYSA-N
SMILES: CCO[Si](CCCN=C=O)(OCC)OCC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results