missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Isoamyl-6-methyl-2-heptyl Myristate 95.0+%, TCI America™
$48.46 - $477.19
Chemical Identifiers
| CAS | 88332-30-3 |
|---|---|
| Molecular Formula | C27H54O2 |
| Molecular Weight (g/mol) | 410.727 |
| InChI Key | JHAGSLRTFDBCLA-UHFFFAOYSA-N |
| Synonym | 3-Isoamyl-6-methyl-2-heptyl Tetradecanoate, 2-Isopentyl-1,5-dimethylhexyl Tetradecanoate, Myristic Acid 3-Isoamyl-6-methyl-2-heptyl Ester, Tetradecanoic Acid 3-Isoamyl-6-methyl-2-heptyl Ester |
| PubChem CID | 44629799 |
| IUPAC Name | [6-methyl-3-(3-methylbutyl)heptan-2-yl] tetradecanoate |
| SMILES | CCCCCCCCCCCCCC(=O)OC(C)C(CCC(C)C)CCC(C)C |
Chemical Identifiers
| 88332-30-3 | |
| 410.727 | |
| 3-Isoamyl-6-methyl-2-heptyl Tetradecanoate, 2-Isopentyl-1,5-dimethylhexyl Tetradecanoate, Myristic Acid 3-Isoamyl-6-methyl-2-heptyl Ester, Tetradecanoic Acid 3-Isoamyl-6-methyl-2-heptyl Ester | |
| [6-methyl-3-(3-methylbutyl)heptan-2-yl] tetradecanoate |
| C27H54O2 | |
| JHAGSLRTFDBCLA-UHFFFAOYSA-N | |
| 44629799 | |
| CCCCCCCCCCCCCC(=O)OC(C)C(CCC(C)C)CCC(C)C |
Specifications
| 88332-30-3 | |
| C27H54O2 | |
| 3-Isoamyl-6-methyl-2-heptyl Tetradecanoate, 2-Isopentyl-1,5-dimethylhexyl Tetradecanoate, Myristic Acid 3-Isoamyl-6-methyl-2-heptyl Ester, Tetradecanoic Acid 3-Isoamyl-6-methyl-2-heptyl Ester | |
| CCCCCCCCCCCCCC(=O)OC(C)C(CCC(C)C)CCC(C)C | |
| 410.727 | |
| 410.73 | |
| Liquid |
| Colorless | |
| 25 g | |
| JHAGSLRTFDBCLA-UHFFFAOYSA-N | |
| [6-methyl-3-(3-methylbutyl)heptan-2-yl] tetradecanoate | |
| 44629799 | |
| ≥95.0% (GC) | |
| 3-Isoamyl-6-methyl-2-heptyl Myristate |
Safety and Handling
EINECSNumber : (2)-0798&(2)-2489
TSCA : No