missing translation for 'onlineSavingsMsg'
Learn More

3-Isoamyl-6-methyl-2-heptyl Myristate 95.0+%, TCI America™

Catalog No. I066225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I066225G 25 g
I0662500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I066225G Supplier TCI America Supplier No. I066225G
Only null left

Chemical Identifiers

CAS 88332-30-3
Molecular Formula C27H54O2
Molecular Weight (g/mol) 410.727
InChI Key JHAGSLRTFDBCLA-UHFFFAOYSA-N
Synonym 3-Isoamyl-6-methyl-2-heptyl Tetradecanoate, 2-Isopentyl-1,5-dimethylhexyl Tetradecanoate, Myristic Acid 3-Isoamyl-6-methyl-2-heptyl Ester, Tetradecanoic Acid 3-Isoamyl-6-methyl-2-heptyl Ester
PubChem CID 44629799
IUPAC Name [6-methyl-3-(3-methylbutyl)heptan-2-yl] tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC(C)C(CCC(C)C)CCC(C)C

Specifications

Color Colorless
Quantity 25 g
Formula Weight 410.73
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Isoamyl-6-methyl-2-heptyl Myristate
EINECSNumber (2)-0798&(2)-2489
TSCA No