missing translation for 'onlineSavingsMsg'
Learn More

3-Iodotoluene 98.0+%, TCI America™

Catalog No. I0216500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0216500G 500 g
I021625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0216500G Supplier TCI America Supplier No. I0216500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 625-95-6
Molecular Formula C7H7I
Molecular Weight (g/mol) 218.037
MDL Number MFCD00001050
InChI Key VLCPISYURGTGLP-UHFFFAOYSA-N
Synonym 3-iodotoluene, m-iodotoluene, benzene, 1-iodo-3-methyl, m-tolyl iodide, m-methyliodobenzene, toluene, m-iodo, 3-methylphenyl iodide, unii-2h574ir35g, 1-iodo-3-methyl-benzene, m-iodo-toluen
PubChem CID 12268
IUPAC Name 1-iodo-3-methylbenzene
SMILES CC1=CC(=CC=C1)I

Specifications

Melting Point -27°C
Boiling Point 213°C
Quantity 500 g
Formula Weight 218.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Iodotoluene
RTECSNumber DA3440000
TSCA Yes